![Naproxen Impurity J certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/207/377/2648fb4e-202f-466e-90d9-202264ad391a/640/2648fb4e-202f-466e-90d9-202264ad391a.png)
Naproxen Impurity J
![Naproxen Impurity J certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/207/377/2648fb4e-202f-466e-90d9-202264ad391a/640/2648fb4e-202f-466e-90d9-202264ad391a.png)
Synonym(s):
2-Ethyl-6-methoxynaphthalene, Ethylnerolin
Empirical Formula (Hill Notation):
C13H14O
CAS Number:
Molecular Weight:
186.25
Products
Brand
Formula Weight
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing