Skip to Content
Merck
Naltrexone hydrochloride European Pharmacopoeia (EP) Reference Standard

Naltrexone hydrochloride

Naltrexone hydrochloride European Pharmacopoeia (EP) Reference Standard
Empirical Formula (Hill Notation):
C20H23NO4 · HCl
CAS Number:
Molecular Weight:
377.86

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Research Area
Application
API Family
Special Grade