![NADH Grade II, disodium salt](/deepweb/assets/sigmaaldrich/product/structures/358/302/6f851c56-072b-44c9-be2a-7474cdb17e76/640/6f851c56-072b-44c9-be2a-7474cdb17e76.png)
NADH
![NADH Grade II, disodium salt](/deepweb/assets/sigmaaldrich/product/structures/358/302/6f851c56-072b-44c9-be2a-7474cdb17e76/640/6f851c56-072b-44c9-be2a-7474cdb17e76.png)
Synonym(s):
β-Nicotinamide adenine dinucleotide, reduced disodium salt hydrate, β-DPNH, β-NADH, DPNH, Diphosphopyridine nucleotide, reduced form, NADH
Empirical Formula (Hill Notation):
C21H27N7Na2O14P2 · xH2O
CAS Number:
Molecular Weight:
709.40 (anhydrous basis)
Products
Formula Weight
Manufacturer
Physical Form
Quality Segment
Showing 1-1 of 1