Skip to Content
Merck
Myristicine phyproof® Reference Substance

Myristicine

Myristicine phyproof® Reference Substance
Synonym(s):
Myristicin, 5-Allyl-2,3-(methylendioxy)anisole, 6-Allyl-4-methoxy-1,3-benzodioxole
Empirical Formula (Hill Notation):
C11H12O3
CAS Number:
Molecular Weight:
192.21

Products

Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing