Multiscreen® 96 well Plate, solid bottom
Synonym(s):
MultiScreen® 96-well plate
Products
Product Category
Brand
Product Line
Binding Type
Wells
Quality Segment
Color
Plate Material
Special Grade
Sterility
Technique
Showing 1-11 of 11
Compare
Product Number
Description
Pricing