Skip to Content
Merck
vibrant-m

Moxifloxacin Related Compound A

vibrant-m
Synonym(s):
1-Cyclopropyl-6,8-difluoro-1,4-dihydro-7-[(4aS,7aS)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-3-quinolinecarboxylic acid
Empirical Formula (Hill Notation):
C20H21F2N3O3
CAS Number:
Molecular Weight:
389.40

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1