![ML349 ≥98% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/277/143/607e8540-5bec-4bda-98b2-403b8de21df5/640/607e8540-5bec-4bda-98b2-403b8de21df5.png)
ML349
![ML349 ≥98% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/277/143/607e8540-5bec-4bda-98b2-403b8de21df5/640/607e8540-5bec-4bda-98b2-403b8de21df5.png)
Synonym(s):
ML349, (5,5-Dioxo-4H-thieno[4,5-c]thiochromen-2-yl)-[4-(4-methoxyphenyl)piperazin-1-yl]methanone
Empirical Formula (Hill Notation):
C23H22N2O4S2
CAS Number:
Molecular Weight:
454.56
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing