Skip to Content
Merck
Methyl benzoate (distilled and stabilized Type A), Technipur®

Methyl benzoate

Methyl benzoate (distilled and stabilized Type A), Technipur®
Synonym(s):
Methyl benzoate
Linear Formula:
C6H5COOCH3
CAS Number:
Molecular Weight:
136.15

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique