![Metconazole PESTANAL®, analytical standard, mixture of stereoisomers](/deepweb/assets/sigmaaldrich/product/structures/158/922/8259893b-9283-499e-b97d-42205394995d/640/8259893b-9283-499e-b97d-42205394995d.png)
Metconazole
![Metconazole PESTANAL®, analytical standard, mixture of stereoisomers](/deepweb/assets/sigmaaldrich/product/structures/158/922/8259893b-9283-499e-b97d-42205394995d/640/8259893b-9283-499e-b97d-42205394995d.png)
Synonym(s):
5-(4-Chlorophenylmethyl)-2,2-dimethyl-1-(1H-1,2,4-triazol-1-ylmethyl)cyclopentanol
Empirical Formula (Hill Notation):
C17H22ClN3O
CAS Number:
Molecular Weight:
319.83
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2