
Koenimbine

Synonym(s):
3,11-Dihydro-8-methoxy-3,3,5-trimethylpyrano[3,2-a]carbazole, 8-Methoxy-3,3,5-trimethyl-3,11-dihydro-chromeno[5,6-b]indole, Kenimbine, Koenimbin, NSC 127152
Empirical Formula (Hill Notation):
C19H19NO2
CAS Number:
Molecular Weight:
293.36
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing