Skip to Content
Merck
Isobutyraldehyde United States Pharmacopeia (USP) Reference Standard

Isobutyraldehyde

Isobutyraldehyde United States Pharmacopeia (USP) Reference Standard
Synonym(s):
2-Methylpropanal, 2-Methylpropionaldehyde, Isobutyraldehyde
Linear Formula:
(CH3)2CHCHO
CAS Number:
Molecular Weight:
72.11

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique