Ipratropium bromide monohydrate
Synonym(s):
Atropine isopropyl bromide
Empirical Formula (Hill Notation):
C20H30BrNO3 Ā· H2O
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing