![Interference Mix 2 (For Qualitative Use Only) 100 μg/μL each analyte in methanol, analytical standard, Cerilliant®](/deepweb/assets/sigmaaldrich/product/images/179/982/660e363d-ae73-4ecb-af4b-b1356c83ee85/640/660e363d-ae73-4ecb-af4b-b1356c83ee85.jpg)
Interference Mix 2 (For Qualitative Use Only)
![Interference Mix 2 (For Qualitative Use Only) 100 μg/μL each analyte in methanol, analytical standard, Cerilliant®](/deepweb/assets/sigmaaldrich/product/images/179/982/660e363d-ae73-4ecb-af4b-b1356c83ee85/640/660e363d-ae73-4ecb-af4b-b1356c83ee85.jpg)
Synonym(s):
Qualitative Interference Mixture
Products
Brand
Manufacturer
Physical Form
Format
Chemical Composition
Application
Package Size
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing