Skip to Content
Merck
vibrant-m

Hemoglobin Protease Substrate

vibrant-m
Synonym(s):
Hemoglobin from bovine blood, Bovine hemoglobin, Hb, Methemoglobin
CAS Number:

Products

Product Category
Manufacturer
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1