Skip to Content
Merck
Guanidinium iodide ≥99%, anhydrous

Guanidinium iodide

Guanidinium iodide ≥99%, anhydrous
Empirical Formula (Hill Notation):
CH6IN3
CAS Number:
Molecular Weight:
186.98

Products

Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing