Skip to Content
Merck
Glyoxal solution for molecular biology, BioReagent, ~40% in H2O (~8.8 M)

Glyoxal solution

Glyoxal solution for molecular biology, BioReagent, ~40% in H2O (~8.8 M)
Synonym(s):
Ethanedial, Oxalaldehyde
Linear Formula:
OHCCHO
CAS Number:
Molecular Weight:
58.04

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2