Ginsenoside Rg2
Synonym(s):
(3β,6α,12β)-3,12,20-Trihydroxydammar-24-en-6-yl 2-O-(6-deoxy-α-L-mannopyranosyl)-β-D-glucopyranoside, Chikusetsusaponin I, Panaxoside Rg2, Prosapogenin C2
Empirical Formula (Hill Notation):
C42H72O13
CAS Number:
Molecular Weight:
785.01
Products
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing