![γ-BHC certified reference material, 1000 μg/mL in methanol](/deepweb/assets/sigmaaldrich/product/images/946/288/62886fe4-c44e-4ebc-bf96-eef4be7a7a74/640/62886fe4-c44e-4ebc-bf96-eef4be7a7a74.jpg)
γ-BHC
![γ-BHC certified reference material, 1000 μg/mL in methanol](/deepweb/assets/sigmaaldrich/product/images/946/288/62886fe4-c44e-4ebc-bf96-eef4be7a7a74/640/62886fe4-c44e-4ebc-bf96-eef4be7a7a74.jpg)
Synonym(s):
Lindane solution, γ-1,2,3,4,5,6-Hexachlorocyclohexane, γ-benzene hexachloride
Empirical Formula (Hill Notation):
C6H6Cl6
CAS Number:
Molecular Weight:
290.83
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing