Skip to Content
Merck
Fmoc-Phe-OH 98%

Fmoc-Phe-OH

Fmoc-Phe-OH 98%
Synonym(s):
N-(9-Fluorenylmethoxycarbonyl)-L-phenylalanine, Fmoc-L-phenylalanine, Fmoc-Phe-OH
Empirical Formula (Hill Notation):
C24H21NO4
CAS Number:
Molecular Weight:
387.43

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing