
Fmoc-Gln-OH

Synonym(s):
Fmoc-Gln-OH
Empirical Formula (Hill Notation):
C20H20N2O5
CAS Number:
Molecular Weight:
368.38
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Functional Group
Application
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1