![Flumethrin PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/403/879/c7e5e250-50ab-44be-9ce9-330e88fb6576/640/c7e5e250-50ab-44be-9ce9-330e88fb6576.png)
Flumethrin
![Flumethrin PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/403/879/c7e5e250-50ab-44be-9ce9-330e88fb6576/640/c7e5e250-50ab-44be-9ce9-330e88fb6576.png)
Synonym(s):
Cyano(4-fluoro-3-phenoxyphenyl)methyl 3-[2-chloro-2-(4-chlorophenyl)ethenyl]-2,2-dimethylcyclopropanecarboxylate
Empirical Formula (Hill Notation):
C28H22Cl2FNO3
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing