Skip to Content
Merck
vibrant-m

Fentanyl for system suitability

vibrant-m
Synonym(s):
Fentanyl
Empirical Formula (Hill Notation):
C22H28N2O
CAS Number:
Molecular Weight:
336.47

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1