![Fenbufen British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/310/922/12ac52ae-fe2d-4e2a-9ae9-6966d3604ab8/640/12ac52ae-fe2d-4e2a-9ae9-6966d3604ab8.png)
Fenbufen
![Fenbufen British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/310/922/12ac52ae-fe2d-4e2a-9ae9-6966d3604ab8/640/12ac52ae-fe2d-4e2a-9ae9-6966d3604ab8.png)
Synonym(s):
γ-Oxo-(1,1′-biphenyl)-4-butanoic acid
Linear Formula:
C6H5C6H4CO(CH2)2CO2H
CAS Number:
Molecular Weight:
254.28
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing