Famphur
Synonym(s):
O-(4-Dimethylsulfamoylphenyl)-O,O-dimethyl-phosphorothioate, Famophos
Empirical Formula (Hill Notation):
C10H16NO5PS2
CAS Number:
Molecular Weight:
325.34
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing