Skip to Content
Merck
Ethyl propionate ≥97%, FCC, FG

Ethyl propionate

Ethyl propionate ≥97%, FCC, FG
Linear Formula:
CH3CH2COOC2H5
CAS Number:
Molecular Weight:
102.13
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing