Skip to Content
Merck
EDDP-D3 perchlorate solution 1.0 mg/mL in methanol (as pyrrolinium), ampule of 1 mL, certified reference material, Cerilliant®

EDDP-D3 perchlorate solution

EDDP-D3 perchlorate solution 1.0 mg/mL in methanol (as pyrrolinium), ampule of 1 mL, certified reference material, Cerilliant®
Synonym(s):
EDDP-D3 perchlorate solution
Empirical Formula (Hill Notation):
C20D3H21N · ClO4
CAS Number:
Molecular Weight:
380.88

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Application
Package Size
Reference Material Type
Special Grade
Technique