
ECOSURF™ EH-9 solution

Synonym(s):
2-Ethyl hexanol EoPo copolymer (9 EO), Alcohol (2EH) alkoxylate, Ethyl hexanol ethoxylated propoxylated
CAS Number:
Products
Product Category
Brand
Sustainability
Feature
Physical Form
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing