Skip to Content
Merck
DL-Norvaline

DL-Norvaline

DL-Norvaline
Synonym(s):
(±)-2-Aminopentanoic acid, DL-Norvaline
Linear Formula:
CH3CH2CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
117.15

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Format
Analyte Suitability
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing