![(±)-Heraclenin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/291/995/cb9e416b-dd37-4751-a546-736f02da8a2d/640/cb9e416b-dd37-4751-a546-736f02da8a2d.png)
(±)-Heraclenin
![(±)-Heraclenin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/291/995/cb9e416b-dd37-4751-a546-736f02da8a2d/640/cb9e416b-dd37-4751-a546-736f02da8a2d.png)
Synonym(s):
Imperatorin epoxide, Oxyimperatorin
Empirical Formula (Hill Notation):
C16H14O5
CAS Number:
Molecular Weight:
286.28
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing