![Diisopentyl phthalate analytical standard](/deepweb/assets/sigmaaldrich/product/structures/257/626/b4e8294d-3dfe-49b3-9c48-ec9433dd4aad/640/b4e8294d-3dfe-49b3-9c48-ec9433dd4aad.png)
Diisopentyl phthalate
![Diisopentyl phthalate analytical standard](/deepweb/assets/sigmaaldrich/product/structures/257/626/b4e8294d-3dfe-49b3-9c48-ec9433dd4aad/640/b4e8294d-3dfe-49b3-9c48-ec9433dd4aad.png)
Synonym(s):
1,2-Benzenedicarboxylic acid 1,2-bis(3-methylbutyl) ester, Bis(3-methylbutyl) phthalate, Diisoamyl phthalate, NSC 17070, Phthalic acid diisopentyl ester
Empirical Formula (Hill Notation):
C18H26O4
CAS Number:
Molecular Weight:
306.40
Products
Product Category
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing