Skip to Content
Merck
Dihydroethidium ≥95%

Dihydroethidium

Dihydroethidium ≥95%
Synonym(s):
2,7-Diamino-10-ethyl-9-phenyl-9,10-dihydrophenanthridine, 3,8-Diamino-5,6-dihydro-5-ethyl-6-phenylphenanthridine, Hydroethidine
Empirical Formula (Hill Notation):
C21H21N3
CAS Number:
Molecular Weight:
315.41

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Showing 1-2 of 2