Dicycloverine hydrochloride
Synonym(s):
Dicyclomine hydrochloride, 2-(Diethylamino)ethyl 1-cyclohexylcyclohexane-1-carboxylate hydrochloride, [1,1-Bicyclohexyl]-1-carboxylic acid 2-(diethylamino)ethyl ester hydrochloride
Empirical Formula (Hill Notation):
C19H35NO2 · HCl
CAS Number:
Molecular Weight:
345.95
Products
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
API Family
Special Grade
Showing 1-2 of 2