![(+)-Fenchone phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/144/652/8839f1f2-6917-4a76-84b8-bce78379f73d/640/8839f1f2-6917-4a76-84b8-bce78379f73d.png)
(+)-Fenchone
![(+)-Fenchone phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/144/652/8839f1f2-6917-4a76-84b8-bce78379f73d/640/8839f1f2-6917-4a76-84b8-bce78379f73d.png)
Synonym(s):
(+)-1,3,3-Trimethyl-2-norbornanone, (1S)-1,3,3-Trimethylbicyclo[2.2.1]heptan-2-one
Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing