![Demeton-S-methyl-sulfon certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/858/955/114fe34c-88d9-4de3-8e17-bfd46d9fd909/640/114fe34c-88d9-4de3-8e17-bfd46d9fd909.png)
Demeton-S-methyl-sulfon
![Demeton-S-methyl-sulfon certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/858/955/114fe34c-88d9-4de3-8e17-bfd46d9fd909/640/114fe34c-88d9-4de3-8e17-bfd46d9fd909.png)
Empirical Formula (Hill Notation):
C6H15O5PS2
CAS Number:
Molecular Weight:
262.28
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing