![Decoquinate British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/186/350/f10cc14f-5019-4582-9bd8-ba73703e597c/640/f10cc14f-5019-4582-9bd8-ba73703e597c.png)
Decoquinate
![Decoquinate British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/186/350/f10cc14f-5019-4582-9bd8-ba73703e597c/640/f10cc14f-5019-4582-9bd8-ba73703e597c.png)
Synonym(s):
Ethyl 6-decyloxy-7-ethoxy-4-hydroxy-3-quinolinecarboxylate
Empirical Formula (Hill Notation):
C24H35NO5
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3