Skip to Content
Merck
Daidzein phyproof® Reference Substance

Daidzein

Daidzein phyproof® Reference Substance
Synonym(s):
4′,7-Dihydroxy­iso­flavone, 7-Hydroxy-3-(4-hydroxy­phenyl)-4H-1-benzo­pyran-4-one, 7-Hydroxy-3-(4-hydroxy­phenyl)­chromone, Isoaurostatin, Daidzein
Empirical Formula (Hill Notation):
C15H10O4
CAS Number:
Molecular Weight:
254.24

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique