![Cycloastragenol phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/160/366/fc63e901-8aba-40e4-86f6-6833b81cc26f/640/fc63e901-8aba-40e4-86f6-6833b81cc26f.png)
Cycloastragenol
![Cycloastragenol phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/160/366/fc63e901-8aba-40e4-86f6-6833b81cc26f/640/fc63e901-8aba-40e4-86f6-6833b81cc26f.png)
Synonym(s):
(3β,6α,16β,20R,24S)-20,24-Epoxy-9,19-cyclolanostane-3,6,16,25-tetrol, Astramembrangenin, Cyclogalegigenin, Cyclosieversigenin
Empirical Formula (Hill Notation):
C30H50O5
CAS Number:
Molecular Weight:
490.72
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing