Skip to Content
Merck
Copper(II) tartrate hydrate ≥95.0% (calc. on dry substance, RT)

Copper(II) tartrate hydrate

Copper(II) tartrate hydrate ≥95.0% (calc. on dry substance, RT)
Synonym(s):
Cupric tartrate hydrate, Tartaric acid cupric salt
Linear Formula:
[-CH(OH)CO2]2Cu·xH2O
CAS Number:
Molecular Weight:
211.62 (anhydrous basis)
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing