Skip to Content
Merck
Closantel PESTANAL®, analytical standard

Closantel

Closantel PESTANAL®, analytical standard
Synonym(s):
5′-Chloro-4′-(4-chloro-α-cyanobenzyl)-3,5-diiodo-2′-methylsalicylanilide, N-[5-Chloro-4-(4-chloro-α-cyanobenzyl)-2-methylphenyl]-2-hydroxy-3,5-diiodobenzamide
Empirical Formula (Hill Notation):
C22H14Cl2I2N2O2
CAS Number:
Molecular Weight:
663.07
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing