![Clorsulon VETRANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/266/183/5de978f8-10b7-45db-afcf-91911dfcf059/640/5de978f8-10b7-45db-afcf-91911dfcf059.png)
Clorsulon
![Clorsulon VETRANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/266/183/5de978f8-10b7-45db-afcf-91911dfcf059/640/5de978f8-10b7-45db-afcf-91911dfcf059.png)
Synonym(s):
4-Amino-6-(trichlorovinyl)-1,3-benzenedisulfonamide
Empirical Formula (Hill Notation):
C8H8Cl3N3O4S2
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing