cis,cis-1,3-Cyclooctadiene
Empirical Formula (Hill Notation):
C8H12
CAS Number:
Molecular Weight:
108.18
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing