Skip to Content
Merck
Cisapride United States Pharmacopeia (USP) Reference Standard

Cisapride

Cisapride United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Cisapride monohydrate, cis-4-Amino-5-chloro-N-[1-[3-(4-fluorophenoxy)propyl]-3-methoxy-4-piperidinyl]-2-methoxybenxamide monohydrate, Acenalin, Cipril
Empirical Formula (Hill Notation):
C23H29N3O4ClF · H2O
CAS Number:
Molecular Weight:
483.96

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing