Skip to Content
Merck
Cerulenin ≥98% (HPLC), from Cephalosporium caerulens

Cerulenin

Cerulenin ≥98% (HPLC), from Cephalosporium caerulens
Synonym(s):
(2R,3S,E,E)-2,3-Epoxy-4-oxo-7,10-dodecadienamide, Cerulenin, Cephalosporium caerulens
Empirical Formula (Hill Notation):
C12H17NO3
CAS Number:
Molecular Weight:
223.27

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Activity Spectrum
Research Area
Quality Segment
Mode of Action