Skip to Content
Merck
Carfentanil Oxalate solution (sold as 100 ug/mL in methanol as free base), certified reference material, ampule of 0.5 mL, Cerilliant®

Carfentanil Oxalate solution

Carfentanil Oxalate solution (sold as 100 ug/mL in methanol as free base), certified reference material, ampule of 0.5 mL, Cerilliant®
Empirical Formula (Hill Notation):
C24H30N2O3 · C2H2O4
CAS Number:
Molecular Weight:
484.54

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique