Skip to Content
Merck
Carcinine dihydrochloride ≥98% (HPLC)

Carcinine dihydrochloride

Carcinine dihydrochloride ≥98% (HPLC)
Synonym(s):
β-Alanylhistamine dihydrochloride, 3-Amino-N-[2-(1H-imidazol-4-yl)ethyl]propanamide dihydrochloride, Alistin, Beta-Alanylhistamine dihydrochloride
Empirical Formula (Hill Notation):
C8H14N4O · 2HCl
CAS Number:
Molecular Weight:
255.14

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing