
Calcipotriol for system suitability

Synonym(s):
Calcipotriol, (5Z,7E,22E,24S)-24-Cyclopropyl-9,10-secochola-5,7,10(19),22-tetraene-1α,3ε,24-triol
Empirical Formula (Hill Notation):
C27H40O3
CAS Number:
Molecular Weight:
412.60
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing