Skip to Content
Merck
Butylated hydroxyanisole certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Butylated hydroxyanisole

Butylated hydroxyanisole certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
2(3)-t-Butyl-4-hydroxyanisole, 2(3)-t-Butylhydroquinone monomethyl ether, BHA, Butylated hydroxyanisole
Linear Formula:
(CH3)3CC6H3(OCH3)OH
CAS Number:
Molecular Weight:
180.24
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique