![Butorphanol tartrate United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/251/476/29303083-a152-4609-b541-faae9b475be9/640/29303083-a152-4609-b541-faae9b475be9.png)
Butorphanol tartrate
![Butorphanol tartrate United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/251/476/29303083-a152-4609-b541-faae9b475be9/640/29303083-a152-4609-b541-faae9b475be9.png)
Synonym(s):
Butorphanol (+)-tartrate salt, L-N-Cyclobutylmethyl-3,14-dihydroxymorphinan (+)-tartrate salt
Empirical Formula (Hill Notation):
C21H29NO2 · C4H6O6
CAS Number:
Molecular Weight:
477.55
Products
Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing