Skip to Content
Merck
Butalbital certified reference material, pharmaceutical secondary standard

Butalbital

Butalbital certified reference material, pharmaceutical secondary standard
Synonym(s):
5-Allyl-5-isobutylbarbituric acid
Empirical Formula (Hill Notation):
C11H16N2O3
CAS Number:
Molecular Weight:
224.26

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Application
Package Size
API Family
Reference Material Type
Special Grade