
Boc-Abu-OH

Synonym(s):
(S)-2-(Boc-amino)butyric acid, Boc-L-2-aminobutyric acid, Boc-Abu-OH
Linear Formula:
C2H5CH[NHCO2C(CH3)3]CO2H
CAS Number:
Molecular Weight:
203.24
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing